siRNA Details
| Gene Symbol: Acmsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134372.1 | NP_599199.1 | 7 | 703 | ||
| GenBank mRNA | AB069781.1 | - | 7 | 703 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acmsd| Gene Symbol: Acmsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134372.1 | NP_599199.1 | 7 | 703 | ||
| GenBank mRNA | AB069781.1 | - | 7 | 703 | ||