siRNA Details
| Gene Symbol: Clstn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134377.1 | NP_599204.1 | 4 | 467 | ||
| XM_008766475.1 | XP_008764697.1 | 3 | 353 | |||
| GenBank mRNA | AJ427342.1 | - | 4 | 467 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clstn2| Gene Symbol: Clstn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134377.1 | NP_599204.1 | 4 | 467 | ||
| XM_008766475.1 | XP_008764697.1 | 3 | 353 | |||
| GenBank mRNA | AJ427342.1 | - | 4 | 467 | ||