siRNA Details
| Gene Symbol: Sulf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134378.2 | NP_599205.1 | 6 | 963 | ||
| XM_008763486.1 | XP_008761708.1 | 6 | 1535 | |||
| XM_008763487.1 | XP_008761709.1 | 6 | 1689 | |||
| XM_008763488.1 | XP_008761710.1 | 6 | 1144 | |||
| GenBank mRNA | AF230072.1 | - | 6 | 964 | ||

Sulf1
Genomic Map