siRNA Details
| Gene Symbol: Elovl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134383.2 | NP_599210.1 | 4 | 397 | ||
| GenBank mRNA | AB071986.1 | - | 4 | 397 | ||
| Gene Symbol: LOC102549542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008761485.1 | XP_008759707.1 | 4 | 540 | ||

Elovl6
Genomic Map