siRNA Details
| Gene Symbol: Gprasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134386.1 | NP_599213.1 | 2 | 3878 | ||
| XM_006257264.2 | XP_006257326.1 | 9 | 2863 | |||
| XM_006257265.2 | XP_006257327.1 | 9 | 3686 | |||
| XM_006257267.2 | XP_006257329.1 | 8 | 3555 | |||
| XM_008773392.1 | XP_008771614.1 | 11 | 3070 | |||
| XM_008773393.1 | XP_008771615.1 | 10 | 2994 | |||
| XM_008773394.1 | XP_008771616.1 | 11 | 3074 | |||
| XM_008773395.1 | XP_008771617.1 | 11 | 3114 | |||
| XM_008773396.1 | XP_008771618.1 | 9 | 3648 | |||
| XM_008773397.1 | XP_008771619.1 | 9 | 2922 | |||
| XM_008773398.1 | XP_008771620.1 | 9 | 2921 | |||
| GenBank mRNA | AB051807.1 | - | 2 | 3878 | ||

Gprasp1
Genomic Map