siRNA Details
| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134398.2 | NP_599225.1 | 5 | 554 | ||
| XM_006243223.2 | XP_006243285.1 | 5 | 586 | |||
| GenBank mRNA | AF178669.1 | - | 5 | 556 | ||
| FQ224819.1 | - | 5 | 529 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aagab| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134398.2 | NP_599225.1 | 5 | 554 | ||
| XM_006243223.2 | XP_006243285.1 | 5 | 586 | |||
| GenBank mRNA | AF178669.1 | - | 5 | 556 | ||
| FQ224819.1 | - | 5 | 529 | |||