siRNA Details
| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134398.2 | NP_599225.1 | 8 | 764 | ||
| XM_006243223.2 | XP_006243285.1 | 8 | 793 | |||
| GenBank mRNA | AF178669.1 | - | 8 | 766 | ||
| FQ224819.1 | - | 8 | 736 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aagab| Gene Symbol: Aagab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134398.2 | NP_599225.1 | 8 | 764 | ||
| XM_006243223.2 | XP_006243285.1 | 8 | 793 | |||
| GenBank mRNA | AF178669.1 | - | 8 | 766 | ||
| FQ224819.1 | - | 8 | 736 | |||