siRNA Details
Gene Symbol: Mk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134399.2 | NP_599226.1 | 1 | 2072 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134399.2 | NP_599226.1 | 1 | 2072 |