siRNA Details
| Gene Symbol: Bzw2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134402.3 | NP_599229.1 | 6 | 664 | ||
| GenBank mRNA | AF031483.1 | - | 6 | 692 | ||
| BC063149.1 | - | 6 | 668 | 6890706 | ||
| FQ225034.1 | - | 6 | 640 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bzw2| Gene Symbol: Bzw2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134402.3 | NP_599229.1 | 6 | 664 | ||
| GenBank mRNA | AF031483.1 | - | 6 | 692 | ||
| BC063149.1 | - | 6 | 668 | 6890706 | ||
| FQ225034.1 | - | 6 | 640 | |||