siRNA Details
Gene Symbol: Yif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172017.2 | NP_742014.2 | 2 | 383 | ||
GenBank mRNA | BC158578.1 | - | 2 | 364 | 7387164 | |
CV074812.1 | - | 2 | 363 | |||
FQ214813.1 | - | 2 | 486 | |||
FQ219390.1 | - | 2 | 378 | |||
FQ220202.1 | - | 2 | 380 | |||
U96490.1 | - | 2 | 324 |

More Information
Other Species Targeted

