siRNA Details
| Gene Symbol: Yif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172017.2 | NP_742014.2 | 4 | 515 | ||
| GenBank mRNA | BC158578.1 | - | 4 | 496 | 7387164 | |
| CV074812.1 | - | 4 | 495 | |||
| FQ219390.1 | - | 4 | 510 | |||
| FQ220202.1 | - | 4 | 512 | |||
| U96490.1 | - | 4 | 456 | |||

Yif1a
Genomic Map