siRNA Details
| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134404.2 | NP_599231.2 | 9 | 980 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Svop| Gene Symbol: Svop | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134404.2 | NP_599231.2 | 9 | 980 | ||