siRNA Details
| Gene Symbol: Slc12a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134405.1 | NP_599232.1 | 7 | 1068 | ||
| GenBank mRNA | AB023645.1 | - | 7 | 1068 | ||
| BC081728.1 | - | 7 | 1098 | 7129648 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc12a9| Gene Symbol: Slc12a9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134405.1 | NP_599232.1 | 7 | 1068 | ||
| GenBank mRNA | AB023645.1 | - | 7 | 1068 | ||
| BC081728.1 | - | 7 | 1098 | 7129648 | ||