siRNA Details
Gene Symbol: Zg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134409.2 | NP_599236.2 | 4 | 338 | ||
GenBank mRNA | AB095177.1 | - | 4 | 338 | ||
CF249177.1 | - | 3 | 310 | |||
Z30584.1 | - | 4 | 364 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zg16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134409.2 | NP_599236.2 | 4 | 338 | ||
GenBank mRNA | AB095177.1 | - | 4 | 338 | ||
CF249177.1 | - | 3 | 310 | |||
Z30584.1 | - | 4 | 364 |