siRNA Details
| Gene Symbol: Ipmk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134417.1 | NP_599244.1 | 6 | 1112 | ||
| XM_006256345.2 | XP_006256407.1 | 6 | 1168 | |||
| XM_006256346.2 | XP_006256408.1 | 6 | 1530 | |||
| XM_008772858.1 | XP_008771080.1 | 6 | 1044 | |||
| GenBank mRNA | AY014898.1 | - | 6 | 1112 | ||

Ipmk
Genomic Map