siRNA Details
| Gene Symbol: Gp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134418.3 | NP_599245.2 | 7, 8 | 1343 | ||
| XM_008759672.1 | XP_008757894.1 | 7, 8 | 1339 | |||
| GenBank mRNA | M58716.1 | - | 7, 8 | 1345 | ||
| X53935.1 | - | 6, 7 | 1244 | |||

Gp2
Genomic Map