siRNA Details
| Gene Symbol: Gp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134418.3 | NP_599245.2 | 3 | 208 | ||
| XM_008759672.1 | XP_008757894.1 | 3 | 204 | |||
| GenBank mRNA | M58716.1 | - | 3 | 210 | ||
| X53935.1 | - | 2 | 109 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gp2| Gene Symbol: Gp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134418.3 | NP_599245.2 | 3 | 208 | ||
| XM_008759672.1 | XP_008757894.1 | 3 | 204 | |||
| GenBank mRNA | M58716.1 | - | 3 | 210 | ||
| X53935.1 | - | 2 | 109 | |||