siRNA Details
Gene Symbol: Hspbap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134419.2 | NP_599246.2 | 5 | 657 | ||
GenBank mRNA | AF168362.1 | - | 5 | 646 | ||
BC091125.1 | - | 5 | 657 | 7366096 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hspbap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134419.2 | NP_599246.2 | 5 | 657 | ||
GenBank mRNA | AF168362.1 | - | 5 | 646 | ||
BC091125.1 | - | 5 | 657 | 7366096 |