siRNA Details
| Gene Symbol: Cd99l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134459.1 | NP_604454.1 | 10 | 1072 | ||
| GenBank mRNA | AB031014.1 | - | 10 | 1072 | ||
| AF481858.1 | - | 10 | 1020 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd99l2| Gene Symbol: Cd99l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134459.1 | NP_604454.1 | 10 | 1072 | ||
| GenBank mRNA | AB031014.1 | - | 10 | 1072 | ||
| AF481858.1 | - | 10 | 1020 | |||