siRNA Details
| Gene Symbol: Capn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134461.1 | NP_604456.1 | 12 | 1748 | ||
| XM_006229729.2 | XP_006229791.2 | 12 | 1985 | |||
| XM_006229730.2 | XP_006229792.1 | 12 | 1877 | |||
| XM_006229731.2 | XP_006229793.1 | 12 | 2121 | |||
| XM_008759673.1 | XP_008757895.1 | 11 | 1912 | |||
| GenBank mRNA | AF484958.1 | - | 12 | 1748 | ||
| FQ223532.1 | - | 12 | 1842 | |||

Capn5
Genomic Map