siRNA Details
Gene Symbol: Capn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134461.1 | NP_604456.1 | 13 | 1858 | ||
GenBank mRNA | AF484958.1 | - | 13 | 1858 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Capn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134461.1 | NP_604456.1 | 13 | 1858 | ||
GenBank mRNA | AF484958.1 | - | 13 | 1858 |