siRNA Details
| Gene Symbol: Crlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134465.2 | NP_604460.2 | 7 | 947 | ||
| XM_008769915.1 | XP_008768137.1 | 7 | 905 | |||
| XM_008769916.1 | XP_008768138.1 | 7 | 930 | |||
| GenBank mRNA | AF404510.1 | - | 7 | 890 | ||
| BC078740.1 | - | 7 | 947 | 7108137 | ||

Crlf2
Genomic Map