siRNA Details
| Gene Symbol: Crlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134465.2 | NP_604460.2 | 4 | 543 | ||
| XM_008769915.1 | XP_008768137.1 | 4 | 501 | |||
| XM_008769916.1 | XP_008768138.1 | 4 | 501 | |||
| GenBank mRNA | AF404510.1 | - | 4 | 486 | ||
| BC078740.1 | - | 4 | 543 | 7108137 | ||
| FQ232217.1 | - | 4 | 529 | |||

Crlf2
Genomic Map