siRNA Details
| Gene Symbol: Gpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138511.1 | NP_612520.1 | 4, 5 | 964 | ||
| GenBank mRNA | L20468.1 | - | 4, 5 | 964 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpc2| Gene Symbol: Gpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138511.1 | NP_612520.1 | 4, 5 | 964 | ||
| GenBank mRNA | L20468.1 | - | 4, 5 | 964 | ||