siRNA Details
| Gene Symbol: Cyp2c22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138512.1 | NP_612521.1 | 7 | 1061 | ||
| GenBank mRNA | EU861213.1 | - | 7 | 1057 | ||
| FQ218628.1 | - | 7 | 1058 | |||
| FQ218929.1 | - | 4 | 575 | |||
| M58041.1 | - | 7 | 1061 | |||
| X53477.1 | - | 7 | 1042 | |||

Cyp2c22
Genomic Map