siRNA Details
| Gene Symbol: Polk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138516.1 | NP_612525.1 | 5 | 687 | ||
| GenBank mRNA | BC166778.1 | - | 5 | 687 | 7934797 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polk| Gene Symbol: Polk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138516.1 | NP_612525.1 | 5 | 687 | ||
| GenBank mRNA | BC166778.1 | - | 5 | 687 | 7934797 | |