siRNA Details
| Gene Symbol: Gzmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138517.3 | NP_612526.2 | 2, 3 | 282 | ||
| GenBank mRNA | BC127475.1 | - | 2, 3 | 262 | 7124652 | |
| FQ220750.1 | - | 2, 3 | 304 | |||
| FQ221957.1 | - | 2, 3 | 304 | |||
| FQ228927.1 | - | 2, 3 | 304 | |||
| Gene Symbol: Gzmbl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002728257.4 | XP_002728303.2 | 2, 3 | 350 | ||

Gzmb
Genomic Map