siRNA Details
| Gene Symbol: Crispld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138518.2 | NP_612527.2 | 5 | 694 | ||
| XM_006255708.2 | XP_006255770.1 | 6 | 817 | |||
| GenBank mRNA | AF109674.1 | - | 5 | 709 | ||
| CB732240.1 | - | 3 | 286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crispld2| Gene Symbol: Crispld2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138518.2 | NP_612527.2 | 5 | 694 | ||
| XM_006255708.2 | XP_006255770.1 | 6 | 817 | |||
| GenBank mRNA | AF109674.1 | - | 5 | 709 | ||
| CB732240.1 | - | 3 | 286 | |||