siRNA Details
| Gene Symbol: Klc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138520.1 | NP_612529.1 | 9 | 1180 | ||
| XM_006228377.2 | XP_006228439.1 | 9 | 1333 | |||
| XM_006228378.2 | XP_006228440.1 | 9 | 1333 | |||
| XM_006228379.2 | XP_006228441.1 | 9 | 1255 | |||
| XM_006228380.2 | XP_006228442.1 | 9 | 1255 | |||
| GenBank mRNA | AF166267.1 | - | 9 | 1180 | ||
| BC078736.1 | - | 9 | 1193 | 7134584 | ||
| BC097284.1 | - | 9 | 1185 | 7454436 | ||

Klc3
Genomic Map