siRNA Details
| Gene Symbol: Cxcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138522.1 | NP_612531.1 | 2 | 141 | ||
| XM_006250721.2 | XP_006250783.1 | 2 | 371 | |||
| GenBank mRNA | D21095.1 | - | 2 | 141 | ||
| D87927.1 | - | 2 | 141 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl3| Gene Symbol: Cxcl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138522.1 | NP_612531.1 | 2 | 141 | ||
| XM_006250721.2 | XP_006250783.1 | 2 | 371 | |||
| GenBank mRNA | D21095.1 | - | 2 | 141 | ||
| D87927.1 | - | 2 | 141 | |||