siRNA Details
| Gene Symbol: Prl5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138527.1 | NP_612536.1 | 5 | 694 | ||
| GenBank mRNA | AB022883.1 | - | 5 | 694 | ||
| AF226607.1 | - | 5 | 661 | |||
| FQ230176.1 | - | 5 | 699 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl5a1| Gene Symbol: Prl5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138527.1 | NP_612536.1 | 5 | 694 | ||
| GenBank mRNA | AB022883.1 | - | 5 | 694 | ||
| AF226607.1 | - | 5 | 661 | |||
| FQ230176.1 | - | 5 | 699 | |||