siRNA Details
| Gene Symbol: Nav2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138529.1 | NP_612538.1 | 31 | 6157 | ||
| XM_006229244.2 | XP_006229306.1 | 33 | 6678 | |||
| XM_008759312.1 | XP_008757534.1 | 32 | 7188 | |||
| XM_008759313.1 | XP_008757535.1 | 31 | 7179 | |||
| XM_008759314.1 | XP_008757536.1 | 31 | 6280 | |||
| XM_008759315.1 | XP_008757537.1 | 27 | 6506 | |||

Nav2
Genomic Map