siRNA Details
Gene Symbol: Nav2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138529.1 | NP_612538.1 | 7 | 1312 | ||
XM_006229244.2 | XP_006229306.1 | 8 | 1824 | |||
XM_008759312.1 | XP_008757534.1 | 7 | 2334 | |||
XM_008759313.1 | XP_008757535.1 | 7 | 2334 | |||
XM_008759314.1 | XP_008757536.1 | 7 | 1435 | |||
XM_008759315.1 | XP_008757537.1 | 2 | 1652 |
