siRNA Details
| Gene Symbol: Nme7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138532.1 | NP_612541.1 | 3 | 275 | ||
| GenBank mRNA | AF202049.1 | - | 3 | 275 | ||
| BC060314.1 | - | 3 | 296 | 6918381 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nme7| Gene Symbol: Nme7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138532.1 | NP_612541.1 | 3 | 275 | ||
| GenBank mRNA | AF202049.1 | - | 3 | 275 | ||
| BC060314.1 | - | 3 | 296 | 6918381 | ||