siRNA Details
| Gene Symbol: Ttl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138536.1 | NP_612545.1 | 5 | 1000 | ||
| XM_006234946.2 | XP_006235008.1 | 5 | 1028 | |||
| GenBank mRNA | AF207605.1 | - | 5 | 1000 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ttl| Gene Symbol: Ttl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138536.1 | NP_612545.1 | 5 | 1000 | ||
| XM_006234946.2 | XP_006235008.1 | 5 | 1028 | |||
| GenBank mRNA | AF207605.1 | - | 5 | 1000 | ||