siRNA Details
Gene Symbol: Nme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138548.2 | NP_612557.1 | 3 | 249 | ||
GenBank mRNA | D13374.1 | - | 3 | 250 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nme1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138548.2 | NP_612557.1 | 3 | 249 | ||
GenBank mRNA | D13374.1 | - | 3 | 250 |