siRNA Details
| Gene Symbol: Prss8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138836.1 | NP_620191.1 | 6 | 1031 | ||
| GenBank mRNA | AB017638.1 | - | 6 | 1031 | ||
| AF202076.1 | - | 6 | 1029 | |||
| BC061800.1 | - | 6 | 1044 | 5598332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prss8| Gene Symbol: Prss8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138836.1 | NP_620191.1 | 6 | 1031 | ||
| GenBank mRNA | AB017638.1 | - | 6 | 1031 | ||
| AF202076.1 | - | 6 | 1029 | |||
| BC061800.1 | - | 6 | 1044 | 5598332 | ||