siRNA Details
| Gene Symbol: Pou3f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138838.1 | NP_620193.1 | 1 | 1536 | ||
| GenBank mRNA | M63712.1 | - | 1 | 1399 | ||
| M72711.1 | - | 1 | 1536 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pou3f1| Gene Symbol: Pou3f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138838.1 | NP_620193.1 | 1 | 1536 | ||
| GenBank mRNA | M63712.1 | - | 1 | 1399 | ||
| M72711.1 | - | 1 | 1536 | |||