siRNA Details
| Gene Symbol: Syngap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113409.2 | NP_001106880.2 | 10 | 1994 | ||
| NM_181092.4 | NP_851606.3 | 12 | 2057 | |||
| GenBank mRNA | AF048976.1 | - | 12 | 2065 | ||
| AF050183.2 | - | 8 | 1746 | |||
| AF058789.2 | - | 12 | 1944 | |||
| AF058790.1 | - | 10 | 1812 | |||

Syngap1
Genomic Map