siRNA Details
| Gene Symbol: Vmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138839.2 | NP_620194.1 | 2 | 175 | ||
| XM_006247078.2 | XP_006247140.1 | 2 | 653 | |||
| XM_006247079.2 | XP_006247141.1 | 3 | 197 | |||
| GenBank mRNA | AF411216.1 | - | 2 | 121 | ||
| BC061721.1 | - | 2 | 175 | 5598561 | ||

Vmp1
Genomic Map