siRNA Details
| Gene Symbol: Unc13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138844.1 | NP_620199.1 | 21 | 2048 | ||
| XM_006247702.2 | XP_006247764.1 | 21 | 2233 | |||
| GenBank mRNA | AF159356.1 | - | 21 | 2048 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Unc13d| Gene Symbol: Unc13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138844.1 | NP_620199.1 | 21 | 2048 | ||
| XM_006247702.2 | XP_006247764.1 | 21 | 2233 | |||
| GenBank mRNA | AF159356.1 | - | 21 | 2048 | ||