siRNA Details
| Gene Symbol: Gpm6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138846.1 | NP_620201.1 | 3 | 610 | ||
| XM_006256849.2 | XP_006256911.1 | 4 | 1149 | |||
| XM_006256850.2 | XP_006256912.1 | 4 | 1151 | |||
| XM_006256851.2 | XP_006256913.1 | 3 | 673 | |||
| XM_008773151.1 | XP_008771373.1 | 3 | 662 | |||
| GenBank mRNA | AB036421.1 | - | 3 | 610 | ||

Gpm6b
Genomic Map