siRNA Details
| Gene Symbol: Rasgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170531.1 | NP_001164002.1 | 24 | 3494 | ||
| GenBank mRNA | BC091317.1 | - | 16 | 2144 | ||
| X67241.1 | - | 24 | 3433 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasgrf1| Gene Symbol: Rasgrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170531.1 | NP_001164002.1 | 24 | 3494 | ||
| GenBank mRNA | BC091317.1 | - | 16 | 2144 | ||
| X67241.1 | - | 24 | 3433 | |||