siRNA Details
| Gene Symbol: Prl2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138861.1 | NP_620216.1 | 4 | 526 | ||
| GenBank mRNA | AF234635.1 | - | 4 | 526 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prl2b1| Gene Symbol: Prl2b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138861.1 | NP_620216.1 | 4 | 526 | ||
| GenBank mRNA | AF234635.1 | - | 4 | 526 | ||