siRNA Details
Gene Symbol: Cand2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181362.1 | NP_852027.1 | 7 | 1005 | ||
XM_006237004.2 | XP_006237066.1 | 9 | 1122 | |||
GenBank mRNA | AB029324.1 | - | 7 | 1005 | ||
AB029341.1 | - | 9 | 1119 | |||
AB029342.1 | - | 6 | 1144 |
