siRNA Details
| Gene Symbol: Ptgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138863.2 | NP_620218.1 | 8 | 711 | ||
| GenBank mRNA | BC089775.1 | - | 8 | 711 | 7307365 | |
| FQ218841.1 | - | 8 | 730 | |||
| FQ220898.1 | - | 8 | 732 | |||
| FQ221589.1 | - | 8 | 727 | |||
| FQ222466.1 | - | 8 | 729 | |||
| FQ229147.1 | - | 8 | 727 | |||
| FQ229679.1 | - | 8 | 727 | |||
| U66322.2 | - | 7 | 676 | |||

Ptgr1
Genomic Map