siRNA Details
| Gene Symbol: Haus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138864.2 | NP_620219.2 | 7 | 738 | ||
| GenBank mRNA | AF092207.1 | - | 734 | |||
| BC127484.1 | - | 7 | 738 | 8363325 | ||
| FQ221448.1 | - | 3 | 252 | |||
| Gene Symbol: NEWGENE_619909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008772211.1 | XP_008770433.1 | 763 | |||

Haus1
Genomic Map