siRNA Details
| Gene Symbol: Hyou1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034028.1 | NP_001029200.1 | 22 | 2588 | ||
| NM_138867.2 | NP_620222.2 | 22 | 2740 | |||
| XM_006242884.2 | XP_006242946.1 | 22 | 2685 | |||
| XM_006242885.2 | XP_006242947.1 | 22 | 2621 | |||
| XM_008766143.1 | XP_008764365.1 | 22 | 2667 | |||
| GenBank mRNA | BC065310.1 | - | 22 | 2631 | 5621638 | |
| U41853.1 | - | 22 | 2740 | |||

Hyou1
Genomic Map