siRNA Details
| Gene Symbol: Golgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138885.1 | NP_620240.1 | 8 | 971 | ||
| XM_006248379.2 | XP_006248441.1 | 9 | 2964 | |||
| XM_006248380.2 | XP_006248442.1 | 8 | 2812 | |||
| XM_008768698.1 | XP_008766920.1 | 8 | 2157 | |||
| XM_008768699.1 | XP_008766921.1 | 8 | 2142 | |||
| XM_008768700.1 | XP_008766922.1 | 8 | 2139 | |||
| XM_008768701.1 | XP_008766923.1 | 7 | 2034 | |||
| XM_008768702.1 | XP_008766924.1 | 6 | 683 | |||
| XM_008768703.1 | XP_008766925.1 | 8 | 2158 | |||
| GenBank mRNA | D25543.1 | - | 8 | 971 | ||

Golgb1
Genomic Map