siRNA Details
| Gene Symbol: Ehd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138890.2 | NP_620245.2 | 6 | 1821 | ||
| XM_006239774.2 | XP_006239836.1 | 7 | 2066 | |||
| XM_008764503.1 | XP_008762725.1 | 6 | 1667 | |||
| GenBank mRNA | AF494093.2 | - | 6 | 1821 | ||

Ehd3
Genomic Map